4-([1,1'-Biphenyl]-3-yl)-6-chloro-2-phenylpyrimidine 97%

Supplier: Ambeed
Warning

A1486655-100MG A1486655-1G A1486655-250MG
76665-222EA 39.26 USD
76665-222 76665-224 76665-226
4-([1,1'-Biphenyl]-3-yl)-6-chloro-2-phenylpyrimidine 97%
4-([1,1'-Biphenyl]-3-yl)-6-chloro-2-phenylpyrimidine

Precaución: for Research Use Only

Formula: C₂₂H₁₅ClN₂
MW: 342.82 g/mol
MDL Number: MFCD32186116
CAS Number: 1852465-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR