(2-(3-Methoxy-3-oxopropyl)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1486556-100MG A1486556-1G A1486556-250MG
76665-202EA 97.61 USD
76665-202 76665-204 76665-206
(2-(3-Methoxy-3-oxopropyl)phenyl)boronic acid 95%
(2-(3-Methoxy-3-oxopropyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₄
MW: 208.0188 g/mol
MDL Number: MFCD28808811
CAS Number: 1001108-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR