(R)-N-((R)-(2-(Di-tert-butylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 95+%

Supplier: Ambeed
Warning

A1486514-100MG A1486514-250MG A1486514-50MG
76665-196EA 183 USD
76665-196 76665-198 76665-200
(R)-N-((R)-(2-(Di-tert-butylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 95+%
(R)-N-((R)-(2-(Di-tert-butylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₂₆H₄₀NOPS
MW: 445.64 g/mol
MDL Number: MFCD33401803
CAS Number: 2757083-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR