(R)-N-((S)-2-(Diphenylphosphino)-1-phenylethyl)-N,2-dimethylpropane-2-sulfinamide 95+%

Supplier: Ambeed
Warning

A1486506-100MG A1486506-250MG A1486506-50MG A1486506-1G
76665-164EA 156 USD
76665-164 76665-166 76665-168 77875-359
(R)-N-((S)-2-(Diphenylphosphino)-1-phenylethyl)-N,2-dimethylpropane-2-sulfinamide 95+%
(R)-N-((S)-2-(Diphenylphosphino)-1-phenylethyl)-N,2-dimethylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₂₅H₃₀NOPS
MDL Number: MFCD33401728
CAS Number: 1824731-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR