(R)-N-((R)-(2-(Di-tert-butylphosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 95+%

Supplier: Ambeed
Warning

A1486505-100MG A1486505-250MG A1486505-50MG
76665-158EA 169 USD
76665-158 76665-160 76665-162
(R)-N-((R)-(2-(Di-tert-butylphosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 95+%
(R)-N-((R)-(2-(Di-tert-butylphosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₂₅H₃₈NOPS
CAS Number: 2565792-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR