(R)-N-((S)-(2-(Di-tert-butylphosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 95%
Supplier: Ambeed
|
Warning
|
A1486504-100MG
A1486504-250MG
A1486504-50MG
76665-152EA
169
USD
76665-152
76665-154
76665-156
(R)-N-((S)-(2-(Di-tert-butylphosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 95%
(R)-N-((S)-(2-(Di-tert-butylphosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide
Precaución: for Research Use Only
Formula:
C₂₅H₃₈NOPS |
MDL Number:
MFCD32697225 CAS Number: 2565792-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...