(R)-N-((R)-(2-(Di(adamantan-1-yl)phosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 95%

Supplier: Ambeed
Warning

A1486502-100MG A1486502-250MG A1486502-50MG
76665-140EA 155.96 USD
76665-140 76665-142 76665-144
(R)-N-((R)-(2-(Di(adamantan-1-yl)phosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 95%
(R)-N-((R)-(2-(Di(adamantan-1-yl)phosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₃₇H₅₀NOPS
MW: 587.8378 g/mol
CAS Number: 2565792-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR