(R)-N-((R)-(2-(Di(adamantan-1-yl)phosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 95+%

Supplier: Ambeed
Warning

A1486501-100MG A1486501-250MG A1486501-50MG
76665-136EA 166 USD
76665-136 76665-138 77875-358
(R)-N-((R)-(2-(Di(adamantan-1-yl)phosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 95+%
(R)-N-((R)-(2-(Di(adamantan-1-yl)phosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₃₈H₅₂NOPS
MDL Number: MFCD32697219
CAS Number: 2509201-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR