(R)-N-((R)-(2-(Dicyclohexylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 95%

Supplier: Ambeed
Warning

A1486499-100MG A1486499-250MG A1486499-50MG
76665-124EA 166.57 USD
76665-124 76665-126 76665-128
(R)-N-((R)-(2-(Dicyclohexylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 95%
(R)-N-((R)-(2-(Dicyclohexylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₃₀H₄₄NOPS
MW: 497.7152 g/mol
MDL Number: MFCD32697197
CAS Number: 2565792-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR