(R)-N-((R)-(2-(Dicyclohexylphosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 95%

Supplier: Ambeed
Warning

A1486498-100MG A1486498-250MG A1486498-50MG
76665-118EA 132 USD
76665-118 76665-120 76665-122
(R)-N-((R)-(2-(Dicyclohexylphosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide 95%
(R)-N-((R)-(2-(Dicyclohexylphosphino)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₂₉H₄₂NOPS
MDL Number: MFCD32697195
CAS Number: 2565792-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR