(R)-N-((R)-(2-(Diphenylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 95+%
Supplier: Ambeed
|
Warning
|
A1486496-100MG
A1486496-250MG
A1486496-50MG
A1486496-1G
76665-106EA
143
USD
76665-106
76665-108
76665-110
77875-356
(R)-N-((R)-(2-(Diphenylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 95+%
(R)-N-((R)-(2-(Diphenylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide
Precaución: for Research Use Only
Formula:
C₃₀H₃₂NOPS |
MDL Number:
MFCD32697152 CAS Number: 2222798-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...