(R)-N-((S)-(2-(Diphenylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 95+%

Supplier: Ambeed
Warning

A1486495-100MG A1486495-250MG A1486495-50MG
76665-100EA 157 USD
76665-100 76665-102 76665-104
(R)-N-((S)-(2-(Diphenylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 95+%
(R)-N-((S)-(2-(Diphenylphosphino)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₃₀H₃₂NOPS
MDL Number: MFCD32697151
CAS Number: 2565792-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR