4-(Dibenzo[b,d]furan-4-yl)aniline 98%
Supplier: Ambeed
|
Warning
|
A1486385-1G
A1486385-25G
A1486385-5G
76665-094EA
11
USD
76665-094
76665-096
76665-098
4-(Dibenzo[b,d]furan-4-yl)aniline 98%
4-(Dibenzo[b,d]furan-4-yl)aniline
Precaución: for Research Use Only
Formula:
C₁₈H₁₃NO |
CAS Number:
578027-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...