2-Bromo-3-fluorotriphenylene 99% (by HPLC)

Supplier: Ambeed
Warning

A1486278-100MG A1486278-1G
76665-092EA 27 USD
76665-092 77875-345
2-Bromo-3-fluorotriphenylene 99% (by HPLC)
2-Bromo-3-fluorotriphenylene

Precaución: for Research Use Only

Formula: C₁₈H₁₀BrF
CAS Number: 2624315-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR