22-Bromodocosanoic acid 98%

Supplier: Ambeed
Warning

A1486236-1G A1486236-250MG A1486236-5G
76665-086EA 111 USD
76665-086 76665-088 76665-090
22-Bromodocosanoic acid 98%
22-Bromodocosanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₄₃BrO₂
CAS Number: 78774-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR