19-Methoxy-19-oxononadecanoic acid 98%

Supplier: Ambeed
Warning

A1486235-1G A1486235-5G
76665-082EA 36.08 USD
76665-082 76665-084
19-Methoxy-19-oxononadecanoic acid 98%
19-Methoxy-19-oxononadecanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₃₈O₄
MW: 342.5133 g/mol
CAS Number: 72849-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR