17-Methoxy-17-oxoheptadecanoic acid 98+%
Supplier: Ambeed
|
Warning
|
A1486234-250MG
A1486234-25G
A1486234-5G
A1486234-1G
76665-076EA
19.1
USD
76665-076
76665-078
76665-080
77597-592
17-Methoxy-17-oxoheptadecanoic acid 98+%
17-Methoxy-17-oxoheptadecanoic acid
Precaución: for Research Use Only
Formula:
C₁₈H₃₄O₄ MW: 314.4602 g/mol |
CAS Number:
72849-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...