2,4-Di-tert-pentylphenol 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Di-tert-amylphenol, 2,4-Bis(1,1-dimethylpropyl)phenol, 2,4-Di-tert-pentylphenol

A1486195-100G A1486195-25G A1486195-500G
76665-058EA 23.34 USD
76665-058 76665-060 76665-062
2,4-Di-tert-pentylphenol 98%
2,4-Di-tert-pentylphenol

Precaución: for Research Use Only

Formula: C₁₆H₂₆O
MW: 234.38 g/mol
MDL Number: MFCD00041929
CAS Number: 120-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR