(R)-Ethyl 2,5-dioxohexahydro-1H-pyrrolizine-7a-carboxylate 97%

Supplier: Ambeed
Warning

A1486119-100MG A1486119-250MG A1486119-1G
76665-046EA 74.27 USD
76665-046 76665-048 77567-092
(R)-Ethyl 2,5-dioxohexahydro-1H-pyrrolizine-7a-carboxylate 97%
(R)-Ethyl 2,5-dioxohexahydro-1H-pyrrolizine-7a-carboxylate

Precaución: For research use only.

Formula: C₁₀H₁₃NO₄
MW: 211.2145 g/mol
CAS Number: 2703745-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR