cis-6-Oxo-7-oxa-5-azaspiro[3.4]octane-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A1486117-100MG A1486117-250MG A1486117-1G
76665-042EA 65 USD
76665-042 76665-044 77624-410
cis-6-Oxo-7-oxa-5-azaspiro[3.4]octane-2-carboxylic acid 97%
cis-6-Oxo-7-oxa-5-azaspiro[3.4]octane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₉NO₄
CAS Number: 2135785-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR