3-(4-Amino-1-oxoisoindolin-2-yl)-1-methylpiperidine-2,6-dione 97%

Supplier: Ambeed
Warning

A1486113-250MG A1486113-1G
76665-040EA 115 USD
76665-040 77567-090
3-(4-Amino-1-oxoisoindolin-2-yl)-1-methylpiperidine-2,6-dione 97%
3-(4-Amino-1-oxoisoindolin-2-yl)-1-methylpiperidine-2,6-dione

Precaución: For research use only.

Formula: C₁₄H₁₅N₃O₃
CAS Number: 2133816-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR