5-Fluoro-2-(1-methyl-2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 97%
Supplier: Ambeed
|
Warning
|
A1486109-1G
A1486109-250MG
A1486109-10G
A1486109-5G
76665-036EA
222
USD
76665-036
76665-038
77875-332
77875-333
5-Fluoro-2-(1-methyl-2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione 97%
5-Fluoro-2-(1-methyl-2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione
Precaución: for Research Use Only
Formula:
C₁₄H₁₁FN₂O₄ |
CAS Number:
2230957-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...