N-(9-((2R,3R,4S,5R)-4-((tert-Butyldimethylsilyl)oxy)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-3H-purin-2-yl)isobutyramide 97%

Supplier: Ambeed
Warning

A1485961-100MG A1485961-250MG
76665-028EA 105 USD
76665-028 76665-030
N-(9-((2R,3R,4S,5R)-4-((tert-Butyldimethylsilyl)oxy)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-3H-purin-2-yl)isobutyramide 97%
N-(9-((2R,3R,4S,5R)-4-((tert-Butyldimethylsilyl)oxy)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-6-oxo-6,9-dihydro-3H-purin-2-yl)isobutyramide

Precaución: for Research Use Only

Formula: C₂₀H₃₃N₅O₆Si
CAS Number: 1237684-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR