3-Methylthiophene-2-carboxaldehyde 85% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-2-thiophenecarboxaldehyde, 3-Methylthiophene-2-carbaldehyde

A1485919-100G A1485919-25G A1485919-5G
76665-014EA 55 USD
76665-014 76665-016 76665-018
3-Methylthiophene-2-carboxaldehyde 85% (by HPLC)
3-Methylthiophene-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₆OS
MDL Number: MFCD00005430
CAS Number: 5834-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR