2-(2,6-Dioxopiperidin-3-yl)-5,6-difluoroisoindoline-1,3-dione 98%

Supplier: Ambeed
Warning

A1485867-100MG A1485867-1G A1485867-5G A1485867-250MG A1485867-25G
76665-010EA 5 USD
76665-010 76665-012 77549-838 77597-582 77624-400
2-(2,6-Dioxopiperidin-3-yl)-5,6-difluoroisoindoline-1,3-dione 98%
2-(2,6-Dioxopiperidin-3-yl)-5,6-difluoroisoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₃H₈F₂N₂O₄
MDL Number: MFCD32853274
CAS Number: 1496997-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR