(2R,3R,4R,5R)-4-(((2S,3R,4S,5R,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal 90%

Supplier: Ambeed
Warning

Synonyms: 2'-Fucosyllactose

A1485771-1G A1485771-5G A1485771-100MG A1485771-250MG
76665-008EA 90.18 USD
76665-008 77567-084 77624-396 77624-398
(2R,3R,4R,5R)-4-(((2S,3R,4S,5R,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal 90%
(2R,3R,4R,5R)-4-(((2S,3R,4S,5R,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal

Precaución: For research use only.

Formula: C₁₈H₃₂O₁₅
MW: 488.44 g/mol
MDL Number: MFCD00079286
CAS Number: 41263-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR