3-(4-Chlorobenzoyl)propionic acid 95%

Supplier: Ambeed
Warning

A1485759-100G A1485759-25G A1485759-5G
76665-002EA 138 USD
76665-002 76665-004 76665-006
3-(4-Chlorobenzoyl)propionic acid 95%
3-(4-Chlorobenzoyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₃
MDL Number: MFCD00002794
CAS Number: 3984-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR