1-Bromo-2,5-difluoro-4-(trifluoromethoxy)benzene 92+%

Supplier: Ambeed
Warning

A1485755-1G A1485755-250MG A1485755-5G
76664-996EA 23.34 USD
76664-996 76664-998 76665-000
1-Bromo-2,5-difluoro-4-(trifluoromethoxy)benzene 92+%
1-Bromo-2,5-difluoro-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₂BrF₅O
MW: 276.9862 g/mol
MDL Number: MFCD25458130
CAS Number: 1439904-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR