3-Bromo-2-chlorobenzotrifluoride 98+%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-chloro-3-(trifluoromethyl)benzene

A1485753-100G A1485753-1G A1485753-25G A1485753-5G
76664-988EA 299.18 USD
76664-988 76664-990 76664-992 76664-994
3-Bromo-2-chlorobenzotrifluoride 98+%
3-Bromo-2-chlorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
MDL Number: MFCD04115994
CAS Number: 56131-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR