α-Bromo-2,5-difluorotoluene 98+%

Supplier: Ambeed
Danger

Synonyms: 2-(Bromomethyl)-1,4-difluorobenzene, 2,5-Difluorobenzyl bromide

A1485749-1G A1485749-25G A1485749-5G
76664-978EA 5.31 USD
76664-978 76664-980 76664-982
α-Bromo-2,5-difluorotoluene 98+%
α-Bromo-2,5-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
MDL Number: MFCD00009897
CAS Number: 85117-99-3
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR