2,4,5-Trifluorobenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2,4,5-trifluorobenzene, α-Bromo-2,4,5-trifluorotoluene

A1485748-25G A1485748-5G
76664-974EA 75.33 USD
76664-974 76664-976
2,4,5-Trifluorobenzyl bromide 98%
2,4,5-Trifluorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
MDL Number: MFCD00061209
CAS Number: 157911-56-3
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR