3-Bromo-4-(trifluoromethoxy)aniline 98+%

Supplier: Ambeed
Warning

A1485746-100G A1485746-25G A1485746-5G
76664-968EA 112.46 USD
76664-968 76664-970 76664-972
3-Bromo-4-(trifluoromethoxy)aniline 98+%
3-Bromo-4-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₇H₅BrF₃NO
MW: 256.02 g/mol
MDL Number: MFCD00190127
CAS Number: 191602-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR