1-Nitro-4-(trifluoromethoxy)benzene 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Trifluoromethoxy)nitrobenzene
A1485742-100G
A1485742-25G
A1485742-5G
76664-954EA
71.09
USD
76664-954
76664-956
76664-958
1-Nitro-4-(trifluoromethoxy)benzene 98+%
1-Nitro-4-(trifluoromethoxy)benzene
Precaución: for Research Use Only
Formula:
C₇H₄F₃NO₃ MW: 207.11 g/mol |
MDL Number:
MFCD00040302 CAS Number: 713-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...