2-Fluoro-6-nitrotoluene 98+%

Supplier: Ambeed
Warning

A1485740-100G A1485740-25G A1485740-5G
76664-948EA 48.81 USD
76664-948 76664-950 76664-952
2-Fluoro-6-nitrotoluene 98+%
2-Fluoro-6-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
MDL Number: MFCD00007197
CAS Number: 769-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR