2-Fluoro-4-nitroanisole 98+%

Supplier: Ambeed
Warning

A1485735-100G A1485735-25G A1485735-5G
76664-924EA 41 USD
76664-924 76664-926 76664-928
2-Fluoro-4-nitroanisole 98+%
2-Fluoro-4-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
MDL Number: MFCD00061095
CAS Number: 455-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR