3-Fluoro-o-xylene 98+%

Supplier: Ambeed
Danger

A1485731-25G A1485731-5G
76664-906EA 62 USD
76664-906 76664-908
3-Fluoro-o-xylene 98+%
3-Fluoro-o-xylene

Precaución: for Research Use Only

Formula: C₈H₉F
MDL Number: MFCD00000323
CAS Number: 443-82-3
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR