3-Chloro-2-nitrotoluene 98+%

Supplier: Ambeed
Warning

A1485730-1G A1485730-25G A1485730-5G
76664-900EA 6.37 USD
76664-900 76664-902 76664-904
3-Chloro-2-nitrotoluene 98+%
3-Chloro-2-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD04117977
CAS Number: 5367-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR