3-Chloro-4-nitrotoluene 98+%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-methyl-1-nitrobenzene

A1485727-100G A1485727-25G A1485727-500G A1485727-5G
76664-884EA 50 USD
76664-884 76664-886 76664-888 76664-890
3-Chloro-4-nitrotoluene 98+%
3-Chloro-4-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MDL Number: MFCD02683043
CAS Number: 38939-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR