3-Amino-4-bromobenzoic acid 98+%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-bromobenzoic Acid

A1485724-100G A1485724-1G A1485724-25G A1485724-5G
76664-876EA 271.6 USD
76664-876 76664-878 76664-880 76664-882
3-Amino-4-bromobenzoic acid 98+%
3-Amino-4-bromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD00579106
CAS Number: 2840-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR