1-Bromo-2-chloro-5-fluorobenzene 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-1-chloro-4-fluorobenzene
A1485721-100G
A1485721-25G
A1485721-500G
A1485721-5G
76664-860EA
56.23
USD
76664-860
76664-862
76664-864
76664-866
1-Bromo-2-chloro-5-fluorobenzene 98+%
1-Bromo-2-chloro-5-fluorobenzene
Precaución: for Research Use Only
Formula:
C₆H₃BrClF MW: 209.44 g/mol |
MDL Number:
MFCD00061120 CAS Number: 201849-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...