(1S,2S)-1,2-Bis(3,5-bis(trifluoromethyl)phenyl)-N1,N2-dimethylethane-1,2-diamine 97%
Supplier: Ambeed
|
Warning
|
A1485602-100MG
A1485602-1G
A1485602-250MG
76664-846EA
93.36
USD
76664-846
76664-848
76664-850
(1S,2S)-1,2-Bis(3,5-bis(trifluoromethyl)phenyl)-N1,N2-dimethylethane-1,2-diamine 97%
(1S,2S)-1,2-Bis(3,5-bis(trifluoromethyl)phenyl)-N1,N2-dimethylethane-1,2-diamine
Precaución: for Research Use Only
Formula:
C₂₀H₁₆F₁₂N₂ MW: 512.3353 g/mol |
CAS Number:
2640520-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...