(3-Acrylamidopropyl)trimethylammonium chloride ~75% in water
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Acrylamido-N,N,N-trimethylpropan-1-aminium chloride
A1483966-100G
A1483966-25G
A1483966-5G
76664-832EA
98.67
USD
76664-832
76664-834
76664-836
(3-Acrylamidopropyl)trimethylammonium chloride ~75% in water
(3-Acrylamidopropyl)trimethylammonium chloride
Precaución: for Research Use Only
Formula:
C₉H₁₉ClN₂O MW: 206.71 g/mol |
MDL Number:
MFCD00274249 CAS Number: 45021-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...