1-Cyclobutylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A1483910-100MG A1483910-250MG
76664-828EA 89.12 USD
76664-828 76664-830
1-Cyclobutylpyrimidine-2,4(1H,3H)-dione 97%
1-Cyclobutylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.17 g/mol
CAS Number: 1797409-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR