6-(Prop-1-yn-1-yl)pyrimidine-2,4(1H,3H)-dione 97%
Supplier: Ambeed
|
Warning
|
A1483909-100MG
A1483909-250MG
76664-824EA
91.24
USD
76664-824
76664-826
6-(Prop-1-yn-1-yl)pyrimidine-2,4(1H,3H)-dione 97%
6-(Prop-1-yn-1-yl)pyrimidine-2,4(1H,3H)-dione
Precaución: for Research Use Only
Formula:
C₇H₆N₂O₂ MW: 150.1347 g/mol |
CAS Number:
1797408-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...