1-(2-Hydroxypropyl)pyrimidine-2,4(1H,3H)-dione 97%
Supplier: Ambeed
|
Warning
|
A1483906-100MG
A1483906-250MG
76664-816EA
51.99
USD
76664-816
76664-818
1-(2-Hydroxypropyl)pyrimidine-2,4(1H,3H)-dione 97%
1-(2-Hydroxypropyl)pyrimidine-2,4(1H,3H)-dione
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂O₃ MW: 170.1659 g/mol |
CAS Number:
1479918-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...