6-Amino-5-ethynylpyrimidin-2(1H)-one 97%

Supplier: Ambeed
Warning

A1483896-100MG A1483896-250MG A1483896-50MG
76664-806EA 132 USD
76664-806 76664-808 76664-810
6-Amino-5-ethynylpyrimidin-2(1H)-one 97%
6-Amino-5-ethynylpyrimidin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₅N₃O
CAS Number: 65223-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR