1-Methyl-6-propylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A1483894-100MG A1483894-1G A1483894-250MG
76664-800EA 29.71 USD
76664-800 76664-802 76664-804
1-Methyl-6-propylpyrimidine-2,4(1H,3H)-dione 97%
1-Methyl-6-propylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.1931 g/mol
CAS Number: 53760-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR