2-(2,4-Dioxo-3,4-dihydropyrimidin-1(2H)-yl)acetamide 97%

Supplier: Ambeed
Warning

A1483893-100MG A1483893-1G A1483893-250MG
76664-794EA 26 USD
76664-794 76664-796 76664-798
2-(2,4-Dioxo-3,4-dihydropyrimidin-1(2H)-yl)acetamide 97%
2-(2,4-Dioxo-3,4-dihydropyrimidin-1(2H)-yl)acetamide

Precaución: for Research Use Only

Formula: C₆H₇N₃O₃
CAS Number: 5358-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR