6-(Dimethylamino)-1-methylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A1483881-100MG A1483881-250MG
76664-776EA 53.05 USD
76664-776 76664-778
6-(Dimethylamino)-1-methylpyrimidine-2,4(1H,3H)-dione 97%
6-(Dimethylamino)-1-methylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MW: 169.1811 g/mol
CAS Number: 1797409-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR