6-Ethyl-1-methylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A1483873-100MG A1483873-1G A1483873-250MG
76664-756EA 28.65 USD
76664-756 76664-758 76664-760
6-Ethyl-1-methylpyrimidine-2,4(1H,3H)-dione 97%
6-Ethyl-1-methylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.1665 g/mol
CAS Number: 1418227-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR